Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DOCK9
Entrez Gene ID
23348
Gene Name
dedicator of cytokinesis 9
Gene Aliases
ZIZ1, ZIZIMIN1
Chromosome Location
Chr.13: 98793487 - 99086406 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130048.1 NP_001123520.1 24 - 25 2851 58
NM_001130049.1 NP_001123521.1 24 - 25 2734 58
NM_001130050.1 NP_001123522.1 24 - 25 2851 58
NM_001318849.1 NP_001305778.1 24 - 25 2734 58
NM_015296.2 NP_056111.1 24 - 25 2734 58
XM_005254034.3 XP_005254091.1 24 - 25 2723 58
XM_005254035.3 XP_005254092.1 24 - 25 2723 58
XM_006719922.1 XP_006719985.1 24 - 25 2884 58
XM_006719924.2 XP_006719987.1 24 - 25 2723 58
XM_006719925.1 XP_006719988.1 24 - 25 2851 58
XM_006719931.3 XP_006719994.1 24 - 25 2679 58
XM_006719932.1 XP_006719995.1 24 - 25 2851 58
XM_006719936.1 XP_006719999.1 24 - 25 2884 58
XM_006719937.3 XP_006720000.1 24 - 25 2679 58
XM_017020503.1 XP_016875992.1 24 - 25 2724 58
XM_017020504.1 XP_016875993.1 24 - 25 2724 58
XM_017020505.1 XP_016875994.1 24 - 25 2724 58
XM_017020506.1 XP_016875995.1 24 - 25 2724 58
XM_017020507.1 XP_016875996.1 24 - 25 2724 58
XM_017020508.1 XP_016875997.1 24 - 25 2724 58
XM_017020509.1 XP_016875998.1 24 - 25 2851 58
XM_017020510.1 XP_016875999.1 24 - 25 2724 58
XM_017020511.1 XP_016876000.1 24 - 25 2724 58
XM_017020512.1 XP_016876001.1 24 - 25 2851 58
XM_017020513.1 XP_016876002.1 24 - 25 2724 58
XM_017020514.1 XP_016876003.1 24 - 25 2851 58
XM_017020515.1 XP_016876004.1 24 - 25 2851 58
XM_017020516.1 XP_016876005.1 24 - 25 2851 58
XM_017020517.1 XP_016876006.1 24 - 25 2823 58
XM_017020518.1 XP_016876007.1 24 - 25 2823 58
XM_017020519.1 XP_016876008.1 24 - 25 2851 58
XM_017020520.1 XP_016876009.1 24 - 25 2851 58
XM_017020521.1 XP_016876010.1 24 - 25 2884 58
GenBank mRNA AB028981.2 - 24 - 25 2757 58
AF527605.1 - 24 - 25 2734 58
AK122987.1 - 13 - 14 1338 58
AK126492.1 - 24 - 25 2851 58
AK127329.1 - 24 - 25 2823 58
AK127508.1 - 22 - 23 2433 58
BC136274.1 - 24 - 25 2714 58
BC171753.1 - 24 - 25 2714 58
CR936820.1 - 24 - 25 2814 58

View Full Product Details