Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DOCK9
Entrez Gene ID
23348
Gene Name
dedicator of cytokinesis 9
Gene Aliases
ZIZ1, ZIZIMIN1
Chromosome Location
Chr.13: 98793487 - 99086406 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130048.1 NP_001123520.1 25 - 26 2913 76
NM_001130049.1 NP_001123521.1 25 - 26 2796 76
NM_001130050.1 NP_001123522.1 25 - 26 2913 76
NM_001318849.1 NP_001305778.1 25 - 26 2796 76
NM_015296.2 NP_056111.1 25 - 26 2796 76
XM_005254034.3 XP_005254091.1 25 - 26 2785 76
XM_005254035.3 XP_005254092.1 25 - 26 2785 76
XM_006719922.1 XP_006719985.1 25 - 26 2946 76
XM_006719924.2 XP_006719987.1 25 - 26 2785 76
XM_006719925.1 XP_006719988.1 25 - 26 2913 76
XM_006719931.3 XP_006719994.1 25 - 26 2741 76
XM_006719932.1 XP_006719995.1 25 - 26 2913 76
XM_006719936.1 XP_006719999.1 25 - 26 2946 76
XM_006719937.3 XP_006720000.1 25 - 26 2741 76
XM_017020503.1 XP_016875992.1 25 - 26 2786 76
XM_017020504.1 XP_016875993.1 25 - 26 2786 76
XM_017020505.1 XP_016875994.1 25 - 26 2786 76
XM_017020506.1 XP_016875995.1 25 - 26 2786 76
XM_017020507.1 XP_016875996.1 25 - 26 2786 76
XM_017020508.1 XP_016875997.1 25 - 26 2786 76
XM_017020509.1 XP_016875998.1 25 - 26 2913 76
XM_017020510.1 XP_016875999.1 25 - 26 2786 76
XM_017020511.1 XP_016876000.1 25 - 26 2786 76
XM_017020512.1 XP_016876001.1 25 - 26 2913 76
XM_017020513.1 XP_016876002.1 25 - 26 2786 76
XM_017020514.1 XP_016876003.1 25 - 26 2913 76
XM_017020515.1 XP_016876004.1 25 - 26 2913 76
XM_017020516.1 XP_016876005.1 25 - 26 2913 76
XM_017020517.1 XP_016876006.1 25 - 26 2885 76
XM_017020518.1 XP_016876007.1 25 - 26 2885 76
XM_017020519.1 XP_016876008.1 25 - 26 2913 76
XM_017020520.1 XP_016876009.1 25 - 26 2913 76
XM_017020521.1 XP_016876010.1 25 - 26 2946 76
GenBank mRNA AB028981.2 - 25 - 26 2819 76
AF527605.1 - 25 - 26 2796 76
AK122987.1 - 14 - 15 1400 76
AK126492.1 - 25 - 26 2913 76
AK127329.1 - 25 - 26 2885 76
BC136274.1 - 25 - 26 2776 76
BC146814.1 - 25 - 26 2790 76
BC171753.1 - 25 - 26 2776 76
CR936820.1 - 25 - 26 2876 76

View Full Product Details