Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NOP16
Entrez Gene ID
51491
Gene Name
NOP16 nucleolar protein
Gene Aliases
HSPC111, HSPC185
Chromosome Location
Chr.5: 176383937 - 176388927 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256539.3 NP_001243468.2 2 - 3 606 66
NM_001256540.3 NP_001243469.2 2 - 3 606 66
NM_001291307.2 NP_001278236.1 2 - 3 606 66
NM_001317975.1 NP_001304904.1 1 - 2 695 66
NM_016391.7 NP_057475.2 2 - 3 606 66
NR_134318.1 2 - 3 606 66
GenBank mRNA AF151875.1 - 3 - 4 674 66
AF161460.1 - 2 - 3 280 66
AK300883.1 - 2 - 3 285 66
AK303281.1 - 2 - 3 277 66
AY561704.1 - 2 - 3 218 66
BC019331.1 - 2 - 3 441 66
BC032424.2 - 2 - 3 249 66
BC040106.1 - 2 - 3 266 66
BP234677.1 - 1 - 2 366 66
BU594763.1 - 2 - 3 280 66
CB122385.1 - 2 - 3 266 66
CR457169.1 - 2 - 3 218 66

View Full Product Details