Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DVL2
Entrez Gene ID
1856
Gene Name
dishevelled segment polarity protein 2
Gene Aliases
-
Chromosome Location
Chr.17: 7225342 - 7234548 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004422.2 NP_004413.1 10 - 11 1385 64
XM_005256502.1 XP_005256559.1 10 - 11 1377 64
XM_017024298.1 XP_016879787.1 7 - 8 731 64
GenBank mRNA AF006012.1 - 10 - 11 1109 64
AK026019.1 - 9 - 10 911 64
AK297099.1 - 10 - 11 1141 64
AK312829.1 - 10 - 11 1386 64
BC014844.1 - 10 - 11 1198 64
BT009822.1 - 10 - 11 1103 64

View Full Product Details