The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193464.1 | NP_001180393.1 | 2 - 3 | 221 | 100 |
NM_015522.3 | NP_056337.1 | 2 - 3 | 221 | 100 | |
NM_016008.3 | NP_057092.2 | 2 - 3 | 221 | 100 | |
XM_005264364.4 | XP_005264421.1 | 2 - 3 | 198 | 100 | |
XM_005264365.4 | XP_005264422.1 | 2 - 3 | 198 | 100 | |
GenBank mRNA | AF151818.1 | - | 2 - 3 | 182 | 100 |
AK074206.1 | - | 2 - 3 | 211 | 100 | |
AK223432.1 | - | 2 - 3 | 221 | 100 | |
AY083346.1 | - | 2 - 3 | 181 | 100 | |
BC006969.1 | - | 2 - 3 | 180 | 100 | |
BC016324.1 | - | 2 - 3 | 215 | 100 | |
BC040558.1 | - | 2 - 3 | 196 | 100 | |
BC058823.1 | - | 2 - 3 | 188 | 100 | |
BG716986.1 | - | 2 - 3 | 226 | 100 |