Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DYNC2LI1
Entrez Gene ID
51626
Gene Name
dynein cytoplasmic 2 light intermediate chain 1
Gene Aliases
CGI-60, D2LIC, LIC3
Chromosome Location
Chr.2: 43774039 - 43828492 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193464.1 NP_001180393.1 8 - 9 750 104
NM_016008.3 NP_057092.2 8 - 9 747 104
XM_005264364.4 XP_005264421.1 8 - 9 727 104
XM_005264365.4 XP_005264422.1 8 - 9 724 104
GenBank mRNA AF151818.1 - 8 - 9 705 104
AK074206.1 - 8 - 9 737 104
AK223432.1 - 8 - 9 747 104
AK301489.1 - 7 - 8 667 104
AL050006.1 - 7 - 8 600 104
AY083346.1 - 8 - 9 707 104
BC006969.1 - 8 - 9 709 104

View Full Product Details