Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DYNC2LI1
Entrez Gene ID
51626
Gene Name
dynein cytoplasmic 2 light intermediate chain 1
Gene Aliases
CGI-60, D2LIC, LIC3
Chromosome Location
Chr.2: 43774039 - 43828492 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193464.1 NP_001180393.1 9 - 10 831 70
NM_016008.3 NP_057092.2 9 - 10 828 70
XM_005264364.4 XP_005264421.1 9 - 10 808 70
XM_005264365.4 XP_005264422.1 9 - 10 805 70
GenBank mRNA AF151818.1 - 9 - 10 786 70
AK074206.1 - 9 - 10 818 70
AK223432.1 - 9 - 10 828 70
AK301489.1 - 8 - 9 748 70
AL050006.1 - 8 - 9 681 70
AY083346.1 - 9 - 10 788 70
BC006969.1 - 9 - 10 790 70

View Full Product Details