Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FASN
Entrez Gene ID
2194
Gene Name
fatty acid synthase
Gene Aliases
FAS, OA-519, SDR27X1
Chromosome Location
Chr.17: 82078338 - 82098303 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004104.4 NP_004095.4 17 - 18 2899 61
XM_011523538.2 XP_011521840.1 17 - 18 2967 61
GenBank mRNA AB209988.1 - 17 - 18 2894 61
AY451392.1 - 16 - 17 2782 61
BC063242.1 - 17 - 18 2899 61
U26644.1 - 16 - 17 2773 61
U29344.1 - 17 - 18 2905 61

View Full Product Details