Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FASN
Entrez Gene ID
2194
Gene Name
fatty acid synthase
Gene Aliases
FAS, OA-519, SDR27X1
Chromosome Location
Chr.17: 82078338 - 82098303 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004104.4 NP_004095.4 27 - 28 4882 82
XM_011523538.2 XP_011521840.1 27 - 28 4950 82
GenBank mRNA AB209988.1 - 27 - 28 4877 82
AY451392.1 - 26 - 27 4765 82
BC014631.2 - 7 - 8 1484 82
BC063242.1 - 27 - 28 4882 82
U26644.1 - 26 - 27 4750 82
U29344.1 - 27 - 28 4882 82

View Full Product Details