Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPTA1
Entrez Gene ID
6708
Gene Name
spectrin alpha, erythrocytic 1
Gene Aliases
EL2, HPP, HS3, SPH3, SPTA
Chromosome Location
Chr.1: 158610498 - 158686716 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003126.2 NP_003117.2 11 - 12 1690 65
XM_011509916.2 XP_011508218.1 12 - 13 1568 65
XM_011509917.2 XP_011508219.1 11 - 12 1604 65
XM_011509918.2 XP_011508220.1 11 - 12 1604 65
XM_011509919.2 XP_011508221.1 11 - 12 1606 65
GenBank mRNA M61877.1 - 11 - 12 1677 65

View Full Product Details