Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRAK1
Entrez Gene ID
22906
Gene Name
trafficking kinesin protein 1
Gene Aliases
MILT1, OIP106
Chromosome Location
Chr.3: 42013802 - 42225890 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042646.2 NP_001036111.1 3 - 4 578 58
NM_001265608.1 NP_001252537.1 3 - 4 578 58
NM_001265609.1 NP_001252538.1 2 - 3 441 58
NM_001265610.1 NP_001252539.1 2 - 3 441 58
NM_014965.4 NP_055780.2 2 - 3 381 58
XM_005264956.2 XP_005265013.1 3 - 4 386 58
XM_005264958.2 XP_005265015.1 2 - 3 441 58
XM_005264960.3 XP_005265017.1 3 - 4 386 58
XM_005264962.3 XP_005265019.1 3 - 4 386 58
XM_005264963.3 XP_005265020.1 3 - 4 386 58
XM_006713029.2 XP_006713092.1 3 - 4 386 58
XM_006713030.2 XP_006713093.1 3 - 4 386 58
XM_006713031.3 XP_006713094.1 3 - 4 386 58
XM_011533489.2 XP_011531791.1 3 - 4 386 58
XM_017005906.1 XP_016861395.1 5 - 6 999 58
XM_017005907.1 XP_016861396.1 2 - 3 363 58
XM_017005908.1 XP_016861397.1 2 - 3 441 58
XM_017005909.1 XP_016861398.1 4 - 5 884 58
XM_017005910.1 XP_016861399.1 2 - 3 441 58
XM_017005911.1 XP_016861400.1 4 - 5 884 58
XM_017005912.1 XP_016861401.1 3 - 4 386 58
XM_017005913.1 XP_016861402.1 3 - 4 386 58
GenBank mRNA AB028965.1 - 3 - 4 578 58
AK000754.1 - 2 - 3 357 58
AK294221.1 - 2 - 3 407 58
AK295480.1 - 3 - 4 616 58
AK295848.1 - 3 - 4 485 58
AK316260.1 - 3 - 4 475 58
AL713787.1 - 2 - 3 345 58
BC015922.1 - 2 - 3 368 58
BX647199.1 - 2 - 3 381 58
DA193280.1 - 2 - 3 442 58

View Full Product Details