Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC25A27
Entrez Gene ID
9481
Gene Name
solute carrier family 25 member 27
Gene Aliases
UCP4
Chromosome Location
Chr.6: 46652915 - 46678190 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_004277.4 NP_004268.3 8 - 9 1172 89
GenBank mRNA AF110532.1 - 8 - 9 894 89
AK090871.1 - 6 - 7 1501 89
AK291427.1 - 8 - 9 1102 89
AK302906.1 - 9 - 10 1212 89
AY358711.1 - 8 - 9 920 89
CB243903.1 - 1 - 2 89

View Full Product Details