The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001190981.1 | NP_001177910.1 | 8 - 9 | 1292 | 87 |
NM_002184.3 | NP_002175.2 | 8 - 9 | 1292 | 87 | |
XM_017009442.1 | XP_016864931.1 | 7 - 8 | 1171 | 87 | |
GenBank mRNA | AB102802.1 | - | 6 - 7 | 979 | 87 |
BC117402.1 | - | 7 - 8 | 1091 | 87 | |
BC117404.1 | - | 7 - 8 | 1091 | 87 | |
HQ628629.1 | - | 6 - 7 | 979 | 87 | |
HQ628630.1 | - | 6 - 7 | 979 | 87 | |
M57230.1 | - | 8 - 9 | 1234 | 87 |