The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005735.1 | NP_001005735.1 | 14 - 15 | 1660 | 108 |
NM_001257387.1 | NP_001244316.1 | 14 - 15 | 1645 | 108 | |
NM_007194.3 | NP_009125.1 | 13 - 14 | 1531 | 108 | |
NM_145862.2 | NP_665861.1 | 12 - 13 | 1444 | 108 | |
XM_006724114.3 | XP_006724177.2 | 9 - 10 | 1174 | 108 | |
XM_006724116.2 | XP_006724179.2 | 9 - 10 | 919 | 108 | |
XM_011529839.2 | XP_011528141.1 | 14 - 15 | 1682 | 108 | |
XM_011529840.2 | XP_011528142.1 | 13 - 14 | 1597 | 108 | |
XM_011529841.1 | XP_011528143.1 | 13 - 14 | 1463 | 108 | |
XM_011529842.2 | XP_011528144.1 | 12 - 13 | 1351 | 108 | |
XM_011529843.1 | XP_011528145.1 | 12 - 13 | 1330 | 108 | |
XM_011529845.2 | XP_011528147.1 | 12 - 13 | 1238 | 108 | |
XM_017028560.1 | XP_016884049.1 | 14 - 15 | 1646 | 108 | |
XM_017028561.1 | XP_016884050.1 | 10 - 11 | 890 | 108 | |
GenBank mRNA | AB040105.1 | - | 11 - 12 | 1372 | 108 |
AF086904.1 | - | 12 - 13 | 1459 | 108 | |
AF096279.1 | - | 12 - 13 | 1459 | 108 | |
AF174135.1 | - | 12 - 13 | 1459 | 108 | |
AF217975.1 | - | 14 - 15 | 2244 | 108 | |
AJ131197.1 | - | 12 - 13 | 1459 | 108 | |
AK289360.1 | - | 13 - 14 | 1510 | 108 | |
AK290754.1 | - | 13 - 14 | 1533 | 108 | |
AW778747.1 | - | 2 - 3 | 108 | ||
AY551295.1 | - | 4 - 5 | 505 | 108 | |
AY551296.1 | - | 5 - 6 | 617 | 108 | |
AY551297.1 | - | 13 - 14 | 1588 | 108 | |
AY551298.1 | - | 11 - 12 | 1397 | 108 | |
AY551299.1 | - | 11 - 12 | 1372 | 108 | |
AY551301.1 | - | 12 - 13 | 1349 | 108 | |
AY551302.1 | - | 11 - 12 | 1368 | 108 | |
AY551303.1 | - | 10 - 11 | 1186 | 108 | |
AY551305.1 | - | 13 - 14 | 1573 | 108 | |
BC004207.2 | - | 13 - 14 | 1486 | 108 | |
CR456418.1 | - | 14 - 15 | 1659 | 108 |