The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005735.1 | NP_001005735.1 | 15 - 16 | 1744 | 71 |
NM_001257387.1 | NP_001244316.1 | 15 - 16 | 1729 | 71 | |
NM_007194.3 | NP_009125.1 | 14 - 15 | 1615 | 71 | |
NM_145862.2 | NP_665861.1 | 13 - 14 | 1528 | 71 | |
XM_006724114.3 | XP_006724177.2 | 10 - 11 | 1258 | 71 | |
XM_006724116.2 | XP_006724179.2 | 10 - 11 | 1003 | 71 | |
XM_011529839.2 | XP_011528141.1 | 15 - 16 | 1766 | 71 | |
XM_011529840.2 | XP_011528142.1 | 14 - 15 | 1681 | 71 | |
XM_011529841.1 | XP_011528143.1 | 14 - 15 | 1547 | 71 | |
XM_011529842.2 | XP_011528144.1 | 13 - 14 | 1435 | 71 | |
XM_011529843.1 | XP_011528145.1 | 13 - 14 | 1414 | 71 | |
XM_011529845.2 | XP_011528147.1 | 13 - 14 | 1322 | 71 | |
XM_017028560.1 | XP_016884049.1 | 15 - 16 | 1730 | 71 | |
XM_017028561.1 | XP_016884050.1 | 11 - 12 | 974 | 71 | |
GenBank mRNA | AB040105.1 | - | 12 - 13 | 1456 | 71 |
AF086904.1 | - | 13 - 14 | 1543 | 71 | |
AF096279.1 | - | 13 - 14 | 1543 | 71 | |
AF174135.1 | - | 13 - 14 | 1543 | 71 | |
AF217975.1 | - | 15 - 16 | 2328 | 71 | |
AJ131197.1 | - | 13 - 14 | 1543 | 71 | |
AK289360.1 | - | 14 - 15 | 1594 | 71 | |
AK290754.1 | - | 14 - 15 | 1617 | 71 | |
AW778747.1 | - | 1 - 2 | 71 | ||
AY551295.1 | - | 5 - 6 | 589 | 71 | |
AY551296.1 | - | 6 - 7 | 701 | 71 | |
AY551297.1 | - | 14 - 15 | 1672 | 71 | |
AY551298.1 | - | 12 - 13 | 1481 | 71 | |
AY551299.1 | - | 12 - 13 | 1456 | 71 | |
AY551300.1 | - | 9 - 10 | 1089 | 71 | |
AY551301.1 | - | 13 - 14 | 1433 | 71 | |
AY551302.1 | - | 12 - 13 | 1452 | 71 | |
AY551303.1 | - | 11 - 12 | 1270 | 71 | |
AY551304.1 | - | 2 - 3 | 400 | 71 | |
AY551305.1 | - | 14 - 15 | 1657 | 71 | |
BC004207.2 | - | 14 - 15 | 1570 | 71 | |
CR456418.1 | - | 15 - 16 | 1743 | 71 |