The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005254034.3 | XP_005254091.1 | 47 - 48 | 5409 | 108 |
XM_005254035.3 | XP_005254092.1 | 47 - 48 | 5316 | 108 | |
XM_017020504.1 | XP_016875993.1 | 48 - 49 | 5479 | 108 | |
XM_017020507.1 | XP_016875996.1 | 47 - 48 | 5410 | 108 | |
XM_017020508.1 | XP_016875997.1 | 48 - 49 | 5386 | 108 | |
XM_017020511.1 | XP_016876000.1 | 47 - 48 | 5341 | 108 | |
XM_017020512.1 | XP_016876001.1 | 48 - 49 | 5606 | 108 | |
XM_017020513.1 | XP_016876002.1 | 47 - 48 | 5317 | 108 | |
XM_017020516.1 | XP_016876005.1 | 47 - 48 | 5537 | 108 | |
XM_017020520.1 | XP_016876009.1 | 47 - 48 | 5444 | 108 | |
GenBank mRNA | AK294573.1 | - | 13 - 14 | 1393 | 108 |
AK300298.1 | - | 12 - 13 | 1422 | 108 | |
AK300331.1 | - | 14 - 15 | 1577 | 108 | |
BC043506.1 | - | 7 - 8 | 716 | 108 | |
BC053620.1 | - | 7 - 8 | 816 | 108 | |
BX648707.1 | - | 10 - 11 | 108 | ||
CR749658.1 | - | 11 - 12 | 2403 | 108 | |
CR936820.1 | - | 46 - 47 | 9780 | 108 |