Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDE8B
Entrez Gene ID
8622
Gene Name
phosphodiesterase 8B
Gene Aliases
ADSD, PPNAD3
Chromosome Location
Chr.5: 77180252 - 77428256 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001029851.2 NP_001025022.1 14 - 15 1665 68
NM_001029852.2 NP_001025023.1 16 - 17 1791 68
NM_001029853.2 NP_001025024.1 16 - 17 1896 68
NM_001029854.2 NP_001025025.1 16 - 17 1815 68
NM_003719.3 NP_003710.1 17 - 18 1956 68
XM_005248621.4 XP_005248678.1 17 - 18 2182 68
XM_005248623.4 XP_005248680.1 17 - 18 1807 68
XM_005248624.4 XP_005248681.1 17 - 18 1751 68
XM_006714725.3 XP_006714788.1 18 - 19 2249 68
XM_006714726.3 XP_006714789.1 18 - 19 2246 68
XM_011543699.2 XP_011542001.1 18 - 19 2251 68
XM_011543700.2 XP_011542002.1 15 - 16 1959 68
XM_011543701.2 XP_011542003.1 18 - 19 1762 68
XM_011543702.2 XP_011542004.1 17 - 18 1593 68
XM_011543703.2 XP_011542005.1 17 - 18 1611 68
XM_011543704.2 XP_011542006.1 15 - 16 1414 68
XM_017010003.1 XP_016865492.1 17 - 18 2184 68
XM_017010004.1 XP_016865493.1 17 - 18 1590 68
XM_017010005.1 XP_016865494.1 14 - 15 1892 68
XM_017010006.1 XP_016865495.1 18 - 19 2029 68
XM_017010007.1 XP_016865496.1 17 - 18 1797 68
XM_017010008.1 XP_016865497.1 14 - 15 1506 68
GenBank mRNA AB085824.1 - 17 - 18 1920 68
AB085825.1 - 14 - 15 1626 68
AB085826.1 - 16 - 17 1752 68
AB085827.1 - 16 - 17 1857 68
AF079529.2 - 17 - 18 1956 68
AK002074.1 - 6 - 7 706 68
AK023913.1 - 12 - 13 1158 68
AL831924.1 - 17 - 18 1786 68
AY129948.1 - 16 - 17 1815 68
AY129949.1 - 14 - 15 1665 68
AY423729.1 - 2 - 3 316 68
BC035144.1 - 4 - 5 412 68

View Full Product Details