The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306212.1 | NP_001293141.1 | 10 - 11 | 1272 | 70 |
NM_001306213.1 | NP_001293142.1 | 10 - 11 | 1324 | 70 | |
NM_001306214.1 | NP_001293143.1 | 9 - 10 | 1266 | 70 | |
NM_003248.5 | NP_003239.2 | 9 - 10 | 1438 | 70 | |
XM_017009798.1 | XP_016865287.1 | 9 - 10 | 1418 | 70 | |
XM_017009799.1 | XP_016865288.1 | 9 - 10 | 1418 | 70 | |
XM_017009800.1 | XP_016865289.1 | 9 - 10 | 1418 | 70 | |
GenBank mRNA | AK302842.1 | - | 9 - 10 | 1266 | 70 |
AK313587.1 | - | 9 - 10 | 1254 | 70 | |
BC050456.1 | - | 10 - 11 | 1522 | 70 | |
Z19585.1 | - | 9 - 10 | 1226 | 70 |