Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANO1
Entrez Gene ID
55107
Gene Name
anoctamin 1
Gene Aliases
DOG1, ORAOV2, TAOS2, TMEM16A
Chromosome Location
Chr.11: 70078302 - 70189545 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018043.5 NP_060513.5 9 - 10 1269 85
NR_030691.1 9 - 10 1117 85
XM_006718602.2 XP_006718665.1 10 - 11 1051 85
XM_006718604.2 XP_006718667.1 10 - 11 1166 85
XM_006718605.2 XP_006718668.1 9 - 10 1112 85
XM_011545121.2 XP_011543423.1 10 - 11 1043 85
XM_011545123.2 XP_011543425.1 10 - 11 1043 85
XM_011545124.2 XP_011543426.1 10 - 11 1043 85
XM_011545125.2 XP_011543427.1 10 - 11 1043 85
XM_011545126.2 XP_011543428.1 10 - 11 1043 85
XM_011545127.2 XP_011543429.1 9 - 10 977 85
XM_011545128.2 XP_011543430.1 9 - 10 977 85
XM_011545129.2 XP_011543431.1 10 - 11 958 85
XM_011545131.2 XP_011543433.1 9 - 10 985 85
XM_017017956.1 XP_016873445.1 9 - 10 977 85
XM_017017957.1 XP_016873446.1 9 - 10 977 85
GenBank mRNA AB845669.1 - 9 - 10 964 85
AK097619.1 - 9 - 10 985 85
AK293088.1 - 9 - 10 977 85
AL833582.1 - 9 - 10 1013 85
AY728143.1 - 9 - 10 1269 85
BC027590.2 - 4 - 5 221 85
BC033036.2 - 9 - 10 1073 85

View Full Product Details