The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290020.1 | NP_001276949.1 | 10 - 11 | 1526 | 90 |
NM_001290021.1 | NP_001276950.1 | 3 - 4 | 352 | 90 | |
NM_001312689.1 | NP_001299618.1 | 5 - 6 | 743 | 90 | |
NM_018076.4 | NP_060546.2 | 10 - 11 | 1479 | 90 | |
XM_011519527.1 | XP_011517829.1 | 10 - 11 | 1526 | 90 | |
XM_011519528.1 | XP_011517830.1 | 10 - 11 | 2176 | 90 | |
XM_011519533.2 | XP_011517835.1 | 5 - 6 | 700 | 90 | |
XM_011519534.2 | XP_011517836.1 | 5 - 6 | 833 | 90 | |
XM_011519535.2 | XP_011517837.1 | 4 - 5 | 585 | 90 | |
XM_017016368.1 | XP_016871857.1 | 10 - 11 | 1526 | 90 | |
XM_017016369.1 | XP_016871858.1 | 10 - 11 | 1526 | 90 | |
XM_017016370.1 | XP_016871859.1 | 10 - 11 | 1526 | 90 | |
XM_017016371.1 | XP_016871860.1 | 10 - 11 | 1526 | 90 | |
GenBank mRNA | AK001679.1 | - | 10 - 11 | 1479 | 90 |
AK292521.1 | - | 10 - 11 | 1482 | 90 | |
AK301708.1 | - | 5 - 6 | 654 | 90 | |
AK302048.1 | - | 3 - 4 | 306 | 90 | |
AK302419.1 | - | 5 - 6 | 1075 | 90 | |
AK302627.1 | - | 5 - 6 | 743 | 90 | |
BC140846.1 | - | 9 - 10 | 1407 | 90 |