The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290020.1 | NP_001276949.1 | 18 - 19 | 2937 | 148 |
NM_001290021.1 | NP_001276950.1 | 11 - 12 | 1763 | 148 | |
NM_001312689.1 | NP_001299618.1 | 13 - 14 | 2154 | 148 | |
NM_018076.4 | NP_060546.2 | 18 - 19 | 2890 | 148 | |
XM_011519527.1 | XP_011517829.1 | 18 - 19 | 2937 | 148 | |
XM_011519528.1 | XP_011517830.1 | 18 - 19 | 3587 | 148 | |
XM_011519533.2 | XP_011517835.1 | 13 - 14 | 2111 | 148 | |
XM_011519534.2 | XP_011517836.1 | 13 - 14 | 2244 | 148 | |
XM_011519535.2 | XP_011517837.1 | 12 - 13 | 1996 | 148 | |
XM_011519537.2 | XP_011517839.1 | 9 - 10 | 1671 | 148 | |
XM_017016368.1 | XP_016871857.1 | 18 - 19 | 2937 | 148 | |
XM_017016369.1 | XP_016871858.1 | 18 - 19 | 2937 | 148 | |
XM_017016370.1 | XP_016871859.1 | 17 - 18 | 2727 | 148 | |
GenBank mRNA | AK292521.1 | - | 18 - 19 | 2893 | 148 |
AK302048.1 | - | 11 - 12 | 1717 | 148 | |
AK302419.1 | - | 13 - 14 | 2486 | 148 | |
AL136859.1 | - | 7 - 8 | 1276 | 148 | |
BC140846.1 | - | 17 - 18 | 2818 | 148 |