Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZBTB46
Entrez Gene ID
140685
Gene Name
zinc finger and BTB domain containing 46
Gene Aliases
BTBD4, BZEL, RINZF, ZNF340, dJ583P15.7, dJ583P15.8
Chromosome Location
Chr.20: 63743668 - 63832332 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025224.3 NP_079500.2 4 - 5 1595 59
XM_005260195.4 XP_005260252.1 4 - 5 1675 59
XM_005260196.3 XP_005260253.1 4 - 5 1491 59
XM_005260197.4 XP_005260254.1 4 - 5 1811 59
XM_005260198.4 XP_005260255.1 4 - 5 1474 59
XM_006723700.3 XP_006723763.1 4 - 5 1507 59
XM_011528548.2 XP_011526850.1 4 - 5 1775 59
GenBank mRNA AK131482.1 - 4 - 5 1595 59
BC052269.1 - 4 - 5 1575 59
BC073800.1 - 4 - 5 1575 59

View Full Product Details