The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080658.1 | NP_542389.1 | 2 - 3 | 155 | 62 |
XM_017018549.1 | XP_016874038.1 | 2 - 3 | 325 | 62 | |
XM_017018550.1 | XP_016874039.1 | 2 - 3 | 359 | 62 | |
GenBank mRNA | AF359506.1 | - | 1 - 2 | 74 | 62 |
AY032593.1 | - | - | 135 | 62 | |
AY040761.1 | - | 2 - 3 | 135 | 62 | |
AY169233.1 | - | 2 - 3 | 155 | 62 | |
BC008689.1 | - | 2 - 3 | 155 | 62 |