Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC14A1
Entrez Gene ID
6563
Gene Name
solute carrier family 14 member 1 (Kidd blood group)
Gene Aliases
HUT11, HsT1341, JK, RACH1, RACH2, UT-B1, UT1, UTE
Chromosome Location
Chr.18: 45724123 - 45752520 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128588.3 NP_001122060.3 2 - 3 212 90
XM_005258329.1 XP_005258386.1 2 - 3 188 90
XM_011526144.1 XP_011524446.1 2 - 3 188 90
GenBank mRNA AK127452.1 - 2 - 3 160 90
AK289608.1 - 2 - 3 155 90
DA186606.1 - 2 - 3 188 90

View Full Product Details