Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CARD9
Entrez Gene ID
64170
Gene Name
caspase recruitment domain family member 9
Gene Aliases
CANDF2, hCARD9
Chromosome Location
Chr.9: 136363956 - 136373681 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052813.4 NP_434700.2 1 - 2 152 98
NM_052814.3 NP_434701.1 1 - 2 152 98
GenBank mRNA AF311287.1 - 1 - 2 113 98
AK024001.1 - 1 - 2 132 98
AK292081.1 - 1 - 2 104 98
AK309043.1 - 1 - 2 117 98
BC008877.2 - 1 - 2 127 98
BI524042.1 - 1 - 2 137 98
BM148244.1 - 1 - 2 152 98

View Full Product Details