Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CARD9
Entrez Gene ID
64170
Gene Name
caspase recruitment domain family member 9
Gene Aliases
CANDF2, hCARD9
Chromosome Location
Chr.9: 136363956 - 136373681 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052813.4 NP_434700.2 8 - 9 1438 95
NM_052814.3 NP_434701.1 8 - 9 1438 95
GenBank mRNA AF311287.1 - 8 - 9 1399 95
AK024001.1 - 8 - 9 1566 95
AK292081.1 - 8 - 9 1390 95
BC008877.2 - 8 - 9 1413 95

View Full Product Details