The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282716.1 | NP_001269645.1 | 1 - 2 | 188 | 69 |
XM_017011685.1 | XP_016867174.1 | 1 - 2 | 349 | 69 | |
GenBank mRNA | AJ007798.2 | - | 1 - 2 | 189 | 69 |