The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282716.1 | NP_001269645.1 | 28 - 29 | 3332 | 62 |
NM_001282717.1 | NP_001269646.1 | 28 - 29 | 3413 | 62 | |
NM_001282718.1 | NP_001269647.1 | 26 - 27 | 3206 | 62 | |
NM_012447.3 | NP_036579.2 | 28 - 29 | 3413 | 62 | |
XM_011515742.1 | XP_011514044.1 | 28 - 29 | 3380 | 62 | |
XM_017011683.1 | XP_016867172.1 | 28 - 29 | 3260 | 62 | |
XM_017011684.1 | XP_016867173.1 | 28 - 29 | 3380 | 62 | |
XM_017011685.1 | XP_016867174.1 | 28 - 29 | 3493 | 62 | |
XM_017011686.1 | XP_016867175.1 | 28 - 29 | 3260 | 62 | |
XM_017011687.1 | XP_016867176.1 | 26 - 27 | 3086 | 62 | |
GenBank mRNA | AK292394.1 | - | 28 - 29 | 3240 | 62 |
AK302587.1 | - | 25 - 26 | 2824 | 62 | |
AK302693.1 | - | 26 - 27 | 3029 | 62 | |
BC140932.1 | - | 28 - 29 | 3172 | 62 | |
BC146806.1 | - | 28 - 29 | 3238 | 62 |