The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002923.3 | NP_002914.1 | 4 - 5 | 475 | 133 |
GenBank mRNA | AF493926.1 | - | 4 - 5 | 442 | 133 |
AK313668.1 | - | 4 - 5 | 475 | 133 | |
AY971351.1 | - | 4 - 5 | 455 | 133 | |
BC007049.1 | - | 4 - 5 | 474 | 133 | |
BT007065.1 | - | 4 - 5 | 442 | 133 | |
CD700013.1 | - | 4 - 5 | 493 | 133 | |
CR457410.1 | - | 4 - 5 | 442 | 133 | |
L13463.1 | - | 4 - 5 | 474 | 133 |