The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079911.2 | NP_001073380.1 | 2 - 3 | 152 | 106 |
NM_004407.3 | NP_004398.1 | 2 - 3 | 152 | 106 | |
XM_011531705.2 | XP_011530007.1 | 2 - 3 | 1027 | 106 | |
XM_011531706.2 | XP_011530008.1 | 2 - 3 | 1027 | 106 | |
GenBank mRNA | BC130581.1 | - | 1 - 2 | 74 | 106 |
BC132865.1 | - | 1 - 2 | 74 | 106 | |
U65378.1 | - | 2 - 3 | 147 | 106 | |
U89012.1 | - | 2 - 3 | 147 | 106 |