Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TESK2
Entrez Gene ID
10420
Gene Name
testis-specific kinase 2
Gene Aliases
-
Chromosome Location
Chr.1: 45343883 - 45491342 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320800.1 NP_001307729.1 7 - 8 891 71
NM_007170.2 NP_009101.2 8 - 9 1199 71
XM_005270355.3 XP_005270412.1 7 - 8 1024 71
XM_006710287.2 XP_006710350.1 9 - 10 1239 71
XM_011540497.1 XP_011538799.1 10 - 11 1272 71
XM_011540500.2 XP_011538802.1 8 - 9 1171 71
XM_017000047.1 XP_016855536.1 8 - 9 1257 71
XM_017000048.1 XP_016855537.1 9 - 10 1527 71
XM_017000049.1 XP_016855538.1 7 - 8 1324 71
GenBank mRNA AB057597.1 - 8 - 9 1191 71
AJ132545.1 - 7 - 8 1143 71
AK027573.1 - 8 - 9 1173 71
AK294172.1 - 7 - 8 866 71
BC022194.1 - 8 - 9 1179 71

View Full Product Details