The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166691.1 | NP_001160163.1 | 2 - 3 | 255 | 180 |
NM_003318.4 | NP_003309.2 | 2 - 3 | 243 | 180 | |
XM_011536099.2 | XP_011534401.1 | 2 - 3 | 217 | 180 | |
XM_011536100.2 | XP_011534402.1 | 2 - 3 | 243 | 180 | |
GenBank mRNA | AK095599.1 | - | 2 - 3 | 715 | 180 |
AK292460.1 | - | 2 - 3 | 218 | 180 | |
AK315696.1 | - | 2 - 3 | 160 | 180 | |
AX775733.1 | - | 2 - 3 | 215 | 180 | |
BC000633.2 | - | 2 - 3 | 206 | 180 | |
BC032858.2 | - | 2 - 3 | 208 | 180 | |
DA853919.1 | - | 2 - 3 | 243 | 180 |