The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003058.3 | NP_003049.2 | 4 - 5 | 1015 | 60 |
GenBank mRNA | AB075951.1 | - | 4 - 5 | 1019 | 60 |
AK290787.1 | - | 4 - 5 | 919 | 60 | |
BC039899.1 | - | 4 - 5 | 896 | 60 | |
X98333.1 | - | 4 - 5 | 985 | 60 |