Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HHIP
Entrez Gene ID
64399
Gene Name
hedgehog interacting protein
Gene Aliases
HIP
Chromosome Location
Chr.4: 144645996 - 144742645 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022475.2 NP_071920.1 5 - 6 1651 78
XM_005263178.4 XP_005263235.1 5 - 6 1520 78
XM_006714288.3 XP_006714351.1 5 - 6 1520 78
GenBank mRNA AK291849.1 - 5 - 6 991 78
AY009317.1 - 5 - 6 1491 78
AY009951.1 - 5 - 6 1626 78
AY358747.1 - 5 - 6 1601 78
BC025311.1 - 5 - 6 1477 78

View Full Product Details