The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024098.3 | NP_077003.1 | 1 - 2 | 933 | 104 |
GenBank mRNA | AK025974.1 | - | 1 - 2 | 819 | 104 |
AK123775.1 | - | 3 - 4 | 606 | 104 | |
AK302329.1 | - | 2 - 3 | 723 | 104 | |
BC001378.1 | - | 1 - 2 | 784 | 104 | |
DQ501251.1 | - | 1 - 2 | 764 | 104 |