The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256369.1 | NP_001243298.1 | 1 - 2 | 158 | 63 |
NM_031200.2 | NP_112477.1 | 1 - 2 | 158 | 63 | |
GenBank mRNA | AF145207.1 | - | 1 - 2 | 101 | 63 |
AF145439.1 | - | 1 - 2 | 135 | 63 | |
AJ132337.1 | - | 1 - 2 | 37 | 63 | |
AK312824.1 | - | 1 - 2 | 113 | 63 | |
AK313366.1 | - | 1 - 2 | 112 | 63 | |
BC069678.1 | - | 1 - 2 | 119 | 63 | |
DB103756.1 | - | 1 - 2 | 158 | 63 |