The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289987.1 | NP_001276916.1 | 3 - 4 | 862 | 81 |
NM_001300866.1 | NP_001287795.1 | 2 - 3 | 696 | 81 | |
NM_015687.3 | NP_056502.1 | 2 - 3 | 696 | 81 | |
XM_005248713.3 | XP_005248770.1 | 2 - 3 | 667 | 81 | |
XM_005248715.4 | XP_005248772.1 | 2 - 3 | 667 | 81 | |
GenBank mRNA | AB086011.1 | - | 1 - 2 | 280 | 81 |
AK097021.1 | - | 2 - 3 | 536 | 81 | |
AK290232.1 | - | 2 - 3 | 408 | 81 | |
AL832009.2 | - | 2 - 3 | 496 | 81 | |
BC136443.1 | - | 2 - 3 | 451 | 81 | |
BC136444.1 | - | 3 - 4 | 617 | 81 | |
BX647178.1 | - | 2 - 3 | 647 | 81 |