Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHEX
Entrez Gene ID
5251
Gene Name
phosphate regulating endopeptidase homolog, X-linked
Gene Aliases
HPDR, HPDR1, HYP, HYP1, LXHR, PEX, XLH
Chromosome Location
Chr.X: 22032327 - 22251310 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000444.5 NP_000435.3 21 - 22 2706 118
XM_011545533.1 XP_011543835.1 17 - 18 1515 118
XM_011545536.2 XP_011543838.1 13 - 14 1548 118
XM_017029579.1 XP_016885068.1 17 - 18 1516 118
GenBank mRNA AD000712.1 - 21 - 22 2303 118
AK301528.1 - 20 - 21 2442 118
AK304551.1 - 14 - 15 1543 118
BC105057.1 - 21 - 22 2228 118
BC105059.1 - 21 - 22 2228 118
BU680665.1 - 1 - 2 118
U75645.1 - 21 - 22 2220 118
U82970.1 - 21 - 22 2747 118
U87284.1 - 21 - 22 2345 118

View Full Product Details