The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015698.5 | NP_056513.2 | 2 - 3 | 407 | 66 |
XM_017029415.1 | XP_016884904.1 | 3 - 4 | 481 | 66 | |
GenBank mRNA | AB209806.1 | - | 2 - 3 | 327 | 66 |
BC000397.2 | - | 2 - 3 | 335 | 66 | |
BC003148.1 | - | 2 - 3 | 335 | 66 | |
BE795098.1 | - | - | 407 | 66 |