The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015698.5 | NP_056513.2 | 7 - 8 | 1096 | 91 |
XM_017029415.1 | XP_016884904.1 | 8 - 9 | 1170 | 91 | |
GenBank mRNA | AB209806.1 | - | 7 - 8 | 1016 | 91 |
BC000397.2 | - | 7 - 8 | 1024 | 91 | |
BC003148.1 | - | 7 - 8 | 1024 | 91 | |
U66359.1 | - | 7 - 8 | 90 |