The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015698.5 | NP_056513.2 | 8 - 9 | 1219 | 72 |
XM_017029415.1 | XP_016884904.1 | 9 - 10 | 1293 | 72 | |
GenBank mRNA | AB209806.1 | - | 8 - 9 | 1139 | 72 |
BC000397.2 | - | 8 - 9 | 1147 | 72 | |
BC003148.1 | - | 8 - 9 | 1147 | 72 | |
U66359.1 | - | 8 - 9 | 1193 | 72 |