Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GMPPB
Entrez Gene ID
29925
Gene Name
GDP-mannose pyrophosphorylase B
Gene Aliases
MDDGA14, MDDGB14, MDDGC14
Chromosome Location
Chr.3: 49721476 - 49723974 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013334.3 NP_037466.2 6 - 7 884 95
NM_021971.2 NP_068806.1 6 - 7 884 95
GenBank mRNA AF135421.1 - 6 - 7 776 95
AK024319.1 - 6 - 7 861 95
AK291700.1 - 6 - 7 859 95
BC001141.1 - 6 - 7 823 95
BC008033.1 - 6 - 7 823 95

View Full Product Details