The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021971.2 | NP_068806.1 | 8 - 9 | 1200 | 76 |
GenBank mRNA | AF135421.1 | - | 8 - 9 | 1092 | 76 |
AK291700.1 | - | 8 - 9 | 1175 | 76 | |
BC001141.1 | - | 8 - 9 | 1139 | 76 | |
BC008033.1 | - | 8 - 9 | 1139 | 76 |