Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ERCC1
Entrez Gene ID
2067
Gene Name
ERCC excision repair 1, endonuclease non-catalytic subunit
Gene Aliases
COFS4, RAD10, UV20
Chromosome Location
Chr.19: 45407333 - 45478866 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001166049.1 NP_001159521.1 5 - 6 670 70
NM_001983.3 NP_001974.1 5 - 6 670 70
NM_202001.2 NP_973730.1 4 - 5 712 70
XM_005258634.1 XP_005258691.1 5 - 6 601 70
XM_005258635.2 XP_005258692.1 5 - 6 1071 70
XM_005258636.4 XP_005258693.1 5 - 6 662 70
XM_005258637.1 XP_005258694.1 5 - 6 601 70
XM_011526610.2 XP_011524912.1 5 - 6 617 70
XM_017026459.1 XP_016881948.1 5 - 6 598 70
XM_017026460.1 XP_016881949.1 5 - 6 1071 70
XM_017026461.1 XP_016881950.1 6 - 7 700 70
XM_017026462.1 XP_016881951.1 6 - 7 665 70
XM_017026463.1 XP_016881952.1 5 - 6 657 70
XM_017026464.1 XP_016881953.1 5 - 6 658 70
XM_017026465.1 XP_016881954.1 5 - 6 1071 70
XM_017026466.1 XP_016881955.1 6 - 7 705 70
GenBank mRNA AB069681.1 - 5 - 6 590 70
AF001925.1 - 5 - 6 666 70
AK092039.1 - 4 - 5 408 70
AK314884.1 - 5 - 6 602 70
BC008930.2 - 5 - 6 586 70
BC052813.1 - 4 - 5 655 70
BM450988.1 - 3 - 4 245 70
BT019806.1 - 4 - 5 524 70
BU627436.1 - 4 - 5 70
M13194.1 - 5 - 6 666 70
M28650.1 - 5 - 6 666 70

View Full Product Details