The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001272095.1 | NP_001259024.1 | 4 - 5 | 827 | 96 |
NM_001272096.1 | NP_001259025.1 | 6 - 7 | 713 | 96 | |
NM_004604.4 | NP_004595.2 | 5 - 6 | 691 | 96 | |
XM_005255521.1 | XP_005255578.1 | 5 - 6 | 608 | 96 | |
GenBank mRNA | AF026007.1 | - | 5 - 6 | 376 | 96 |
AF318489.1 | - | 4 - 5 | 750 | 96 | |
AJ000541.1 | - | 4 - 5 | 512 | 96 | |
AK091833.1 | - | 6 - 7 | 713 | 96 | |
AK296652.1 | - | 4 - 5 | 747 | 96 | |
AK315716.1 | - | 5 - 6 | 646 | 96 | |
BC002436.2 | - | 5 - 6 | 600 | 96 | |
BT007326.1 | - | 5 - 6 | 376 | 96 | |
CR541806.1 | - | 5 - 6 | 376 | 96 | |
U07158.1 | - | 5 - 6 | 442 | 96 | |
X85784.1 | - | 5 - 6 | 376 | 96 |