The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318736.1 | NP_001305665.1 | 6 - 7 | 1341 | 137 |
NM_024725.3 | NP_079001.2 | 5 - 6 | 1206 | 137 | |
XM_005274292.3 | XP_005274349.1 | 3 - 4 | 1086 | 137 | |
XM_006718911.3 | XP_006718974.1 | 6 - 7 | 1158 | 137 | |
XM_011542984.2 | XP_011541286.1 | 5 - 6 | 1870 | 137 | |
XM_011542987.2 | XP_011541289.1 | 6 - 7 | 1302 | 137 | |
XM_011542988.2 | XP_011541290.1 | 4 - 5 | 1178 | 137 | |
XM_017018305.1 | XP_016873794.1 | 4 - 5 | 2951 | 137 | |
XM_017018306.1 | XP_016873795.1 | 5 - 6 | 3035 | 137 | |
XM_017018307.1 | XP_016873796.1 | 6 - 7 | 1677 | 137 | |
XM_017018308.1 | XP_016873797.1 | 3 - 4 | 2911 | 137 | |
XM_017018309.1 | XP_016873798.1 | 3 - 4 | 1046 | 137 | |
XM_017018310.1 | XP_016873799.1 | 3 - 4 | 1087 | 137 | |
XM_017018311.1 | XP_016873800.1 | 5 - 6 | 2882 | 137 | |
XM_017018312.1 | XP_016873801.1 | 6 - 7 | 1801 | 137 | |
GenBank mRNA | AF245436.1 | - | 5 - 6 | 1194 | 137 |
AK027171.1 | - | 2 - 3 | 411 | 137 | |
AK056790.1 | - | 5 - 6 | 137 | ||
AK074306.1 | - | 6 - 7 | 136 | ||
BC033726.2 | - | 2 - 3 | 137 |