The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000921.4 | NP_000912.3 | 5 - 6 | 1578 | 76 |
NM_001244683.1 | NP_001231612.1 | 4 - 5 | 835 | 76 | |
XM_017019420.1 | XP_016874909.1 | 5 - 6 | 1559 | 76 | |
XM_017019421.1 | XP_016874910.1 | 5 - 6 | 4276 | 76 | |
GenBank mRNA | AJ005036.1 | - | 5 - 6 | 1560 | 76 |
AK308317.1 | - | 4 - 5 | 835 | 76 | |
BC117369.1 | - | 5 - 6 | 1578 | 76 | |
BC117371.1 | - | 5 - 6 | 1578 | 76 | |
M91667.2 | - | 5 - 6 | 1567 | 76 | |
U36798.1 | - | 5 - 6 | 1538 | 76 |