The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015139.2 | NP_055954.1 | 1 - 2 | 586 | 74 |
XM_006710478.1 | XP_006710541.1 | 1 - 2 | 586 | 74 | |
XM_011541070.1 | XP_011539372.1 | 1 - 2 | 586 | 74 | |
GenBank mRNA | AB044343.1 | - | 1 - 2 | 234 | 74 |
AK289800.1 | - | 1 - 2 | 584 | 74 | |
BC093786.1 | - | 1 - 2 | 228 | 74 | |
BC112031.1 | - | 1 - 2 | 240 | 74 | |
D87449.1 | - | 1 - 2 | 288 | 74 |