The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000831.3 | NP_000822.2 | 15 - 16 | 2701 | 89 |
GenBank mRNA | AW898512.1 | - | 1 - 2 | 47 | 89 |
U16127.1 | - | 15 - 16 | 2584 | 89 |