The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000831.3 | NP_000822.2 | 7 - 8 | 1242 | 84 |
GenBank mRNA | AJ249210.1 | - | 7 - 8 | 1128 | 84 |
AJ299451.1 | - | 7 - 8 | 1107 | 84 | |
U16127.1 | - | 7 - 8 | 1125 | 84 |